missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-5-fluoroaniline, 97%

CAS: 4863-91-6 | C6H5ClFN | 145.56 g/mol

$400.38 - $400.38

Chemical Identifiers

CAS 4863-91-6
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD03407962
InChI Key LPIFAHAICWJMRR-UHFFFAOYSA-N
Synonym pubchem2953, acmc-209kdj, 3-chloro-5-fluorobenzenamine, 5-chloro-3-fluorophenylamine, ksc493m9f, 3-chloro-5-fluoro-phenylamine, buttpark 44\01-96
PubChem CID 2734838
IUPAC Name 3-chloro-5-fluoroaniline
SMILES C1=C(C=C(C=C1F)Cl)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC374740050
SDS View Documents
Thermo Scientific Chemicals
374740050
5 g Glass bottle
Each for $400.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4863-91-6
145.56
LPIFAHAICWJMRR-UHFFFAOYSA-N
2734838
C1=C(C=C(C=C1F)Cl)N
C6H5ClFN
MFCD03407962
pubchem2953, acmc-209kdj, 3-chloro-5-fluorobenzenamine, 5-chloro-3-fluorophenylamine, ksc493m9f, 3-chloro-5-fluoro-phenylamine, buttpark 44\01-96
3-chloro-5-fluoroaniline

Specifications

4863-91-6
96.0
1.4500g/mL
Authentic
Liquid
96% min. (GC)
C6H5ClFN
MFCD03407962
Solubility in water: soluble
C1=C(C=C(C=C1F)Cl)N
145.56
145.56
3-chloro-5-fluoroaniline
Orange to Yellow
100.0
5 g
Glass bottle
1.5610 to 1.563
1.45
ClC6H3(F)NH2
pubchem2953, acmc-209kdj, 3-chloro-5-fluorobenzenamine, 5-chloro-3-fluorophenylamine, ksc493m9f, 3-chloro-5-fluoro-phenylamine, buttpark 44\01-96
LPIFAHAICWJMRR-UHFFFAOYSA-N
3-chloro-5-fluoroaniline
2734838
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

Recommended Storage : Darkens on exposure to light and air

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only