missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4-pyridinecarboxaldehyde 98.0+%, TCI America™
$59.65 - $233.00
Chemical Identifiers
| CAS | 72990-37-5 |
|---|---|
| Molecular Formula | C6H4ClNO |
| Molecular Weight (g/mol) | 141.554 |
| MDL Number | MFCD06200712 |
| InChI Key | ZVGDKOQPJCOCLI-UHFFFAOYSA-N |
| Synonym | 3-chloroisonicotinaldehyde, 3-chloropyridine-4-carboxaldehyde, 3-chloro-4-pyridinecarboxaldehyde, 3-chloro-4-pyridinecarbaldehyde, 3-chloro-pyridine-4-carbaldehyde, 3-chloro-4-formylpyridine, 4-pyridinecarboxaldehyde, 3-chloro, acmc-1bgrb, ksc377a6f |
| PubChem CID | 2762995 |
| IUPAC Name | 3-chloropyridine-4-carbaldehyde |
| SMILES | C1=CN=CC(=C1C=O)Cl |
Chemical Identifiers
| 72990-37-5 | |
| 141.554 | |
| ZVGDKOQPJCOCLI-UHFFFAOYSA-N | |
| 2762995 | |
| C1=CN=CC(=C1C=O)Cl |
| C6H4ClNO | |
| MFCD06200712 | |
| 3-chloroisonicotinaldehyde, 3-chloropyridine-4-carboxaldehyde, 3-chloro-4-pyridinecarboxaldehyde, 3-chloro-4-pyridinecarbaldehyde, 3-chloro-pyridine-4-carbaldehyde, 3-chloro-4-formylpyridine, 4-pyridinecarboxaldehyde, 3-chloro, acmc-1bgrb, ksc377a6f | |
| 3-chloropyridine-4-carbaldehyde |
Specifications
| 72990-37-5 | |
| Yellow | |
| C6H4ClNO | |
| 1 g | |
| ZVGDKOQPJCOCLI-UHFFFAOYSA-N | |
| 3-chloropyridine-4-carbaldehyde | |
| 2762995 | |
| ≥98.0% (GC) | |
| 3-Chloro-4-pyridinecarboxaldehyde |
| 60°C | |
| 93°C | |
| MFCD06200712 | |
| 3-chloroisonicotinaldehyde, 3-chloropyridine-4-carboxaldehyde, 3-chloro-4-pyridinecarboxaldehyde, 3-chloro-4-pyridinecarbaldehyde, 3-chloro-pyridine-4-carbaldehyde, 3-chloro-4-formylpyridine, 4-pyridinecarboxaldehyde, 3-chloro, acmc-1bgrb, ksc377a6f | |
| C1=CN=CC(=C1C=O)Cl | |
| 141.554 | |
| 141.55 | |
| Crystalline Powder |
Safety and Handling
TSCA : No