missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4-pyridinecarboxaldehyde 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 60°C |
| Color | Yellow |
| Boiling Point | 93°C |
| Quantity | 5 g |
| Formula Weight | 141.55 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Chloro-4-pyridinecarboxaldehyde |
Safety and Handling
| TSCA | No |