missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-pyridinecarboxaldehyde 98.0+%, TCI America™

Catalog No. C25421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25421G 1 g
C25425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25421G Supplier TCI America Supplier No. C25421G

Chemical Identifiers

CAS 72990-37-5
Molecular Formula C6H4ClNO
Molecular Weight (g/mol) 141.554
MDL Number MFCD06200712
InChI Key ZVGDKOQPJCOCLI-UHFFFAOYSA-N
Synonym 3-chloroisonicotinaldehyde, 3-chloropyridine-4-carboxaldehyde, 3-chloro-4-pyridinecarboxaldehyde, 3-chloro-4-pyridinecarbaldehyde, 3-chloro-pyridine-4-carbaldehyde, 3-chloro-4-formylpyridine, 4-pyridinecarboxaldehyde, 3-chloro, acmc-1bgrb, ksc377a6f
PubChem CID 2762995
IUPAC Name 3-chloropyridine-4-carbaldehyde
SMILES C1=CN=CC(=C1C=O)Cl

Specifications

Melting Point 60°C
Color Yellow
Boiling Point 93°C
Quantity 1 g
Formula Weight 141.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-pyridinecarboxaldehyde