missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-pyridinecarboxaldehyde, 97%

Catalog No. AC426550050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426550050 5 g Glass bottle
AC426550010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC426550050 Supplier Thermo Scientific Chemicals Supplier No. 426550050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 72990-37-5 | C6H4ClNO | 141.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 72990-37-5
Molecular Formula C6H4ClNO
Molecular Weight (g/mol) 141.56
MDL Number MFCD06200712
InChI Key ZVGDKOQPJCOCLI-UHFFFAOYSA-N
Synonym 3-chloroisonicotinaldehyde, 3-chloropyridine-4-carboxaldehyde, 3-chloro-4-pyridinecarboxaldehyde, 3-chloro-4-pyridinecarbaldehyde, 3-chloro-pyridine-4-carbaldehyde, 3-chloro-4-formylpyridine, 4-pyridinecarboxaldehyde, 3-chloro, acmc-1bgrb, ksc377a6f
PubChem CID 2762995
IUPAC Name 3-chloropyridine-4-carbaldehyde
SMILES C1=CN=CC(=C1C=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 67°C to 68°C
Color White to Yellow
Boiling Point 225°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 141.56
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-pyridinecarboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only