missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-morpholino-1,2,5-thiadiazole 98.0+%, TCI America™

Catalog No. C21895G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21895G 5 g
C218925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21895G Supplier TCI America Supplier No. C21895G
Only null left

Chemical Identifiers

CAS 30165-96-9
Molecular Formula C6H8ClN3OS
Molecular Weight (g/mol) 205.66
MDL Number MFCD00274160
InChI Key LAUCCQWGVCJGFT-UHFFFAOYSA-N
PubChem CID 121645
IUPAC Name 4-(4-chloro-1,2,5-thiadiazol-3-yl)morpholine
SMILES C1COCCN1C2=NSN=C2Cl

Specifications

Melting Point 46°C
Color Yellow
Quantity 5 g
Formula Weight 205.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-morpholino-1,2,5-thiadiazole