missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-hydroxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. C120325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C120325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C120325G Supplier TCI America Supplier No. C120325G
Only null left

Chemical Identifiers

CAS 33697-81-3
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00004349
InChI Key IYTUKSIOQKTZEG-UHFFFAOYSA-N
PubChem CID 118534
ChEBI CHEBI:47106
IUPAC Name 2-(3-chloro-4-hydroxyphenyl)acetic acid
SMILES C1=CC(=C(C=C1CC(=O)O)Cl)O

Specifications

Melting Point 109°C
Quantity 25 g
Formula Weight 186.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-hydroxyphenylacetic Acid
RTECSNumber CY1576300
TSCA No