missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-hydroxybenzaldehyde 97.0+%, TCI America™

Catalog No. p-7125319
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21835G 5 g
C218325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21835G Supplier TCI America Supplier No. C21835G

Chemical Identifiers

CAS 2420-16-8
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.57
MDL Number MFCD00016981
InChI Key VGSOCYWCRMXQAB-UHFFFAOYSA-N
Synonym benzaldehyde, 3-chloro-4-hydroxy, 4-hydroxy-3-chlorobenzaldehyde, 3-chloro-4-hydroxy-benzaldehyde, benzaldehyde, 3-chloro-4-hydroxy-8ci 9ci, pubchem7764, acmc-1crqk, 3-chlor-4-hydroxybenzaldehyde, 5-chloro-4-hyroxybenzaldehyde, 5-chloro-4-hydroxybenzaldehyde, 3-chloro-4-hydroxy benzaldehyde
PubChem CID 17022
IUPAC Name 3-chloro-4-hydroxybenzaldehyde
SMILES OC1=CC=C(C=O)C=C1Cl

Specifications

Melting Point 133°C
Boiling Point 150°C
Quantity 5 g
Formula Weight 156.57
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-hydroxybenzaldehyde