missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4'-fluoropropiophenone 96.0+%, TCI America™
$80.85 - $223.33
Chemical Identifiers
| CAS | 347-93-3 |
|---|---|
| Molecular Formula | C9H8ClFO |
| Molecular Weight (g/mol) | 186.61 |
| MDL Number | MFCD00000991 |
| InChI Key | AAHQPLJUSLMHHR-UHFFFAOYSA-N |
| Synonym | 3-chloro-4'-fluoropropiophenone, 3-chloro-1-4-fluorophenyl propan-1-one, 3-chloro-1-4-fluorophenyl-1-propanone, 3-chloro-1-4-fluorophenyl-1-propanone, 1-propanone, 3-chloro-1-4-fluorophenyl, a-chloropropiophenone, a-chloropropiophenone, pubchem13250, ksc573a4j, ksc573a4j, 3-chloro-4'fluoropropiophenone |
| PubChem CID | 67672 |
| IUPAC Name | 3-chloro-1-(4-fluorophenyl)propan-1-one |
| SMILES | C1=CC(=CC=C1C(=O)CCCl)F |
Chemical Identifiers
| 347-93-3 | |
| 186.61 | |
| AAHQPLJUSLMHHR-UHFFFAOYSA-N | |
| 67672 | |
| C1=CC(=CC=C1C(=O)CCCl)F |
| C9H8ClFO | |
| MFCD00000991 | |
| 3-chloro-4'-fluoropropiophenone, 3-chloro-1-4-fluorophenyl propan-1-one, 3-chloro-1-4-fluorophenyl-1-propanone, 3-chloro-1-4-fluorophenyl-1-propanone, 1-propanone, 3-chloro-1-4-fluorophenyl, a-chloropropiophenone, a-chloropropiophenone, pubchem13250, ksc573a4j, ksc573a4j, 3-chloro-4'fluoropropiophenone | |
| 3-chloro-1-(4-fluorophenyl)propan-1-one |
Specifications
| 347-93-3 | |
| 103°C | |
| MFCD00000991 | |
| 3-chloro-4'-fluoropropiophenone, 3-chloro-1-4-fluorophenyl propan-1-one, 3-chloro-1-4-fluorophenyl propan-1-one, 3-chloro-1-4-fluorophenyl-1-propanone, p-fluoro-, p-fluoro-, pubchem13250, pubchem13250, ksc573a4j, ksc573a4j, 3-chloro-4'fluoropropiophenone | |
| C1=CC(=CC=C1C(=O)CCCl)F | |
| 186.61 | |
| 186.61 | |
| Crystalline Powder |
| 47°C | |
| C9H8ClFO | |
| 5 g | |
| AAHQPLJUSLMHHR-UHFFFAOYSA-N | |
| 3-chloro-1-(4-fluorophenyl)propan-1-one | |
| 67672 | |
| ≥96.0% (GC) | |
| 3-Chloro-4′-fluoropropiophenone |
Safety and Handling
TSCA : No