missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluoroaniline 98.0+%, TCI America™

Catalog No. p-7124748
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
C102425G 25 g
C1024500G 500 g
2 options

Catalog No. C102425G

Supplier: TCI America C102425G

Only null left

Chemical Identifiers

CAS 367-21-5
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD00007767
InChI Key YSEMCVGMNUUNRK-UHFFFAOYSA-N
Synonym benzenamine, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenamine, 3-chloro-4-fluoro-phenylamine, aniline, 3-chloro-4-fluoro, 3-chloro-4-fluoro aniline, unii-7l63cc70uq, 3-chloro-4-fluorophenylamine, 4-fluoro-3-chloroaniline, 3-chloro-4-fluoro-aniline, 3-chlor-4-fluoranilin
PubChem CID 9708
IUPAC Name 3-chloro-4-fluoroaniline
SMILES NC1=CC=C(F)C(Cl)=C1

Specifications

Melting Point 45°C
Color Black-Purple
Quantity 25 g
Physical Form Crystal-Powder at 20°C
UN Number 2811
Formula Weight 145.56
Percent Purity ≥98.0% (GC)
Chemical Name or Material 3-Chloro-4-fluoroaniline
EINECSNumber (3)-3998
TSCA Yes