missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloro-4'-fluoroacetophenone 98.0+%, TCI America™

Catalog No. p-7125934
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33465G 5 g
C334625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33465G Supplier TCI America Supplier No. C33465G

Chemical Identifiers

CAS 2923-66-2
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.58
MDL Number MFCD00042203
InChI Key PCJPESKRPOTNGU-UHFFFAOYSA-N
Synonym 3'-chloro-4'-fluoroacetophenone, 3-chloro-4-fluoroacetophenone, 1-3-chloro-4-fluorophenyl ethanone, 1-3-chloro-4-fluorophenyl ethan-1-one, ethanone, 1-3-chloro-4-fluorophenyl, 1-3-chloro-4-fluoro-phenyl-ethanone, 1-acetyl-3-chloro-4-fluorobenzene, pubchem4202, ksc699k7b, 3-chloro-4-fluoro acetophenone
PubChem CID 520422
IUPAC Name 1-(3-chloro-4-fluorophenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(F)C(Cl)=C1

Specifications

Melting Point 43°C
Boiling Point 127°C
Quantity 5 g
Formula Weight 172.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Chloro-4′-fluoroacetophenone