missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4-ethoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$61.51 - $61.51
Chemical Identifiers
| CAS | 279261-81-3 |
|---|---|
| Molecular Formula | C8H10BClO3 |
| MDL Number | MFCD03701556 |
| InChI Key | QZMQZKMQXURQNC-UHFFFAOYSA-N |
| PubChem CID | 3717658 |
| IUPAC Name | (3-chloro-4-ethoxyphenyl)boronic acid |
Chemical Identifiers
| 279261-81-3 | |
| MFCD03701556 | |
| 3717658 |
| C8H10BClO3 | |
| QZMQZKMQXURQNC-UHFFFAOYSA-N | |
| (3-chloro-4-ethoxyphenyl)boronic acid |
Specifications
| 279261-81-3 | |
| White-Yellow | |
| MFCD03701556 | |
| 3-chloro-4-ethoxyphenyl boronic acid, 3-chloro-4-ethoxybenzeneboronic acid, 3-chloro-4-ethoxypheylboronic acid, 3-chloro-4-ethoxy-phenyl boronic acid, acmc-209gzs, 3-chloro-4-ethoxyphenyl boronicacid, 3-chloranyl-4-ethoxy-phenyl boronic acid, boronic acid,b-3-chloro-4-ethoxyphenyl, 3-chloro-4-ethoxyphenylboronicacid containsvaryingamountsofanhydride | |
| B(C1=CC(=C(C=C1)OCC)Cl)(O)O | |
| 200.425 | |
| 200.43 | |
| 3-Chloro-4-ethoxyphenylboronic Acid (contains varying amounts of Anhydride) |
| 239°C | |
| C8H10BClO3 | |
| 1 g | |
| QZMQZKMQXURQNC-UHFFFAOYSA-N | |
| (3-chloro-4-ethoxyphenyl)boronic acid | |
| 3717658 | |
| Crystalline Powder |
Safety and Handling
TSCA : No