missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(3-fluorobenzyloxy)aniline 98.0+%, TCI America™

Catalog No. C28741G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28741G 1 g
C28745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28741G Supplier TCI America Supplier No. C28741G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 202197-26-0
Molecular Formula C13H11ClFNO
Molecular Weight (g/mol) 251.69
MDL Number MFCD06809822
InChI Key AYPFEYDGZDPAPE-UHFFFAOYSA-N
PubChem CID 7059263
IUPAC Name 3-chloro-4-[(3-fluorophenyl)methoxy]aniline
SMILES NC1=CC(Cl)=C(OCC2=CC(F)=CC=C2)C=C1

Specifications

Melting Point 80°C
Quantity 1 g
Physical Form Crystal-Powder at 20°C
UN Number 3077
Formula Weight 251.69
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 3-Chloro-4-(3-fluorobenzyloxy)aniline