missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4-(2-pyridylmethoxy)aniline 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 93°C |
| Quantity | 5 g |
| Physical Form | Crystal-Powder at 20°C |
| Formula Weight | 234.68 |
| Percent Purity | ≥98.0% (GC,T) |
| Chemical Name or Material | 3-Chloro-4-(2-pyridylmethoxy)aniline |
Safety and Handling
| TSCA | No |