missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(2-pyridylmethoxy)aniline 98.0+%, TCI America™

Catalog No. C31501G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31501G 1 g
C31505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31501G Supplier TCI America Supplier No. C31501G
Only null left

Chemical Identifiers

CAS 524955-09-7
Molecular Formula C12H11ClN2O
Molecular Weight (g/mol) 234.68
MDL Number MFCD08699389
InChI Key XCAPJQSICQSUJP-UHFFFAOYSA-N
PubChem CID 16781212
IUPAC Name 3-chloro-4-[(pyridin-2-yl)methoxy]aniline
SMILES NC1=CC(Cl)=C(OCC2=CC=CC=N2)C=C1

Specifications

Melting Point 93°C
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Formula Weight 234.68
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 3-Chloro-4-(2-pyridylmethoxy)aniline