missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methoxypyridine 97.0+%, TCI America™

Catalog No. C25651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25651G 1 g
C25655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25651G Supplier TCI America Supplier No. C25651G
Only null left

Chemical Identifiers

CAS 13472-84-9
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD03095227
InChI Key MSICGKUDCSUMJZ-UHFFFAOYSA-N
Synonym 3-chloro-2-methoxy-pyridine, pyridine, 3-chloro-2-methoxy, 2-methoxy-3-chloropyridine, pubchem1189, acmc-1c0pw, ksc494g7h, 3-chloranyl-2-methoxy-pyridine
PubChem CID 13295749
IUPAC Name 3-chloro-2-methoxypyridine
SMILES COC1=C(C=CC=N1)Cl

Specifications

Color Yellow
Boiling Point 53°C
Quantity 1 g
UN Number 1993
Formula Weight 143.57
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-methoxypyridine