missing translation for 'onlineSavingsMsg'
Learn More

(3-Chloro-2-hydroxypropyl)trimethylammonium Chloride (ca. 65% in Water), TCI America™

Catalog No. p-7124840
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C118025ML 25 mL
C1180500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C118025ML Supplier TCI America Supplier No. C118025ML
Only null left

Chemical Identifiers

CAS 3327-22-8
Molecular Formula C6H15Cl2NO
Molecular Weight (g/mol) 188.092
MDL Number MFCD00055655
InChI Key CSPHGSFZFWKVDL-UHFFFAOYSA-M
PubChem CID 18732
IUPAC Name (3-chloro-2-hydroxypropyl)-trimethylazanium;chloride
SMILES C[N+](C)(C)CC(CCl)O.[Cl-]

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 188.09
Physical Form Liquid
Chemical Name or Material (3-Chloro-2-hydroxypropyl)trimethylammonium Chloride (ca. 65% in Water)
EINECSNumber (2)-0181
RTECSNumber BP5275400
TSCA Yes