missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Chloro-2-hydroxypropyl)trimethylammonium Chloride (ca. 65% in Water), TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 500 mL |
| Formula Weight | 188.09 |
| Physical Form | Liquid |
| Chemical Name or Material | (3-Chloro-2-hydroxypropyl)trimethylammonium Chloride (ca. 65% in Water) |
Safety and Handling
| EINECSNumber | (2)-0181 |
| RTECSNumber | BP5275400 |
| TSCA | Yes |