missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-hydroxypropyl Methacrylate (stabilized with HQ) 83.0+%, TCI America™

Catalog No. p-7125286
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C210525G 25 g
C2105500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C210525G Supplier TCI America Supplier No. C210525G
Only null left

Chemical Identifiers

CAS 13159-52-9
Molecular Formula C7H11ClO3
Molecular Weight (g/mol) 178.612
MDL Number MFCD00040686
InChI Key DDKMFQGAZVMXQV-UHFFFAOYSA-N
Synonym Methacrylic Acid 3-Chloro-2-hydroxypropyl Ester
PubChem CID 114470
IUPAC Name (3-chloro-2-hydroxypropyl) 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(CCl)O

Specifications

Color Yellow
Boiling Point 108°C
Quantity 25 g
Formula Weight 178.61
Percent Purity ≥83.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-hydroxypropyl Methacrylate (stabilized with HQ)
EINECSNumber (2)-1032
TSCA Yes
Recommended Storage Refrigerator