missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-2-hydroxypropyl Methacrylate (stabilized with HQ) 83.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 108°C |
| Quantity | 500 g |
| Formula Weight | 178.61 |
| Percent Purity | ≥83.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Chloro-2-hydroxypropyl Methacrylate (stabilized with HQ) |
Safety and Handling
| EINECSNumber | (2)-1032 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |