missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-hydrazino-5-(trifluoromethyl)pyridine, 97%

Catalog No. p-7032405
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3385003 1 g
AAH3385006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3385003 Supplier Thermo Scientific Chemicals Supplier No. H3385003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 89570-82-1 | C6H5ClF3N3 | 211.572 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89570-82-1
Molecular Formula C6H5ClF3N3
Molecular Weight (g/mol) 211.572
MDL Number MFCD00067865
InChI Key KHFKSHYCVQZAQP-UHFFFAOYSA-N
Synonym 3-chloro-2-hydrazinyl-5-trifluoromethyl pyridine, 3-chloro-5-trifluoromethyl pyrid-2-ylhydrazine, 3-chloro-2-hydrazino-5-trifluoromethyl pyridine, 1-3-chloro-5-trifluoromethyl pyridin-2-yl hydrazine, 3-chloro-5-trifluoromethyl pyridin-2-ylhydrazine, 3-chloro-5-trifluoromethyl pyridin-2-yl hydrazine, pyridine,3-chloro-2-hydrazinyl-5-trifluoromethyl, 1-3-chloro-5-trifluoromethyl-2-pyridyl hydrazine, 3-chloro-5-trifluoromethyl-2-pyridylhydrazine
PubChem CID 1272691
IUPAC Name [3-chloro-5-(trifluoromethyl)pyridin-2-yl]hydrazine
SMILES C1=C(C=NC(=C1Cl)NN)C(F)(F)F

Specifications

Melting Point 92°C to 94°C
Quantity 1 g
UN Number UN2811
Formula Weight 211.6
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-hydrazino-5-(trifluoromethyl)pyridine
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only