missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-ethoxypyridine 98.0+%, TCI America™

Catalog No. C30815G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30815G 5 g
C30811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30815G Supplier TCI America Supplier No. C30815G
Only null left

Chemical Identifiers

CAS 177743-06-5
Molecular Formula C7H8ClNO
Molecular Weight (g/mol) 157.597
MDL Number MFCD09702462
InChI Key MTTKGSCSPHCOOF-UHFFFAOYSA-N
PubChem CID 11959087
IUPAC Name 3-chloro-2-ethoxypyridine
SMILES CCOC1=C(C=CC=N1)Cl

Specifications

Color Yellow
Quantity 5 g
Formula Weight 157.60
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-ethoxypyridine