missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine 98.0+%, TCI America™

Catalog No. p-7125191
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18665G 5 g
C186625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18665G Supplier TCI America Supplier No. C18665G
Only null left

Chemical Identifiers

CAS 32943-25-2
Molecular Formula C14H12ClN
Molecular Weight (g/mol) 229.707
MDL Number MFCD01632171
InChI Key MHUXTOYYIDFXRF-UHFFFAOYSA-N
PubChem CID 118364
IUPAC Name 2-chloro-6,11-dihydro-5H-benzo[b][1]benzazepine
SMILES C1CC2=CC=CC=C2NC3=C1C=CC(=C3)Cl

Specifications

Melting Point 87°C
Quantity 5 g
Synonym 4-Chloro-2,2′C-iminodibenzyl
Formula Weight 229.71
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine
EINECSNumber (5)-1096
TSCA No