missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1-phenyl-1-propanol 98.0+%, TCI America™

Catalog No. C29045G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29045G 5 g
C290425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29045G Supplier TCI America Supplier No. C29045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18776-12-0
Molecular Formula C9H11ClO
Molecular Weight (g/mol) 170.636
MDL Number MFCD09878800
InChI Key JZFUHAGLMZWKTF-UHFFFAOYSA-N
Synonym alpha-(2-Chloroethyl)benzyl Alcohol
PubChem CID 572064
IUPAC Name 3-chloro-1-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)C(CCCl)O

Specifications

Melting Point 32°C
Color White-Yellow
Boiling Point 116°C
Quantity 5 g
Formula Weight 170.64
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-1-phenyl-1-propanol
TSCA No
Recommended Storage Refrigerator