missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1-butene 98.0+%, TCI America™

Catalog No. p-7124510
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C058125G 25 g
C0581100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C058125G Supplier TCI America Supplier No. C058125G

Chemical Identifiers

CAS 563-52-0
Molecular Formula C4H7Cl
Molecular Weight (g/mol) 90.55
MDL Number MFCD00000869
InChI Key VZGLVCFVUREVDP-UHFFFAOYSA-N
Synonym 3-chloro-1-butene, 1-butene, 3-chloro, 1-methylallyl chloride, alpha-methallyl chloride, alpha-methylallyl chloride, .alpha.-methallyl chloride, gamma-chloro-alpha-butylene, 2-chloro-3-butene, .alpha.-methylallyl chloride, .gamma.-chloro-.alpha.-butylene
PubChem CID 11242
IUPAC Name 3-chlorobut-1-ene
SMILES CC(C=C)Cl

Specifications

Color Colorless
Boiling Point 64°C
Quantity 25 g
UN Number 1993
Formula Weight 90.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-1-butene
EINECSNumber (2)-0117
RTECSNumber EM4261000
TSCA Yes
Recommended Storage Refrigerator