missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxybenzaldehyde, 98%, Thermo Scientific™

Catalog No. AC296560050
Change view
Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296560050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC296560050 Supplier Thermo Scientific Chemicals Supplier No. 296560050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 619-21-6
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.13
MDL Number MFCD00039575
InChI Key UHDNUPHSDMOGCR-UHFFFAOYSA-N
Synonym 3-carboxybenzaldehyde, benzoic acid, 3-formyl, m-formyl benzoic acid, 3-formyl benzoic acid, 3-formyl-benzoic acid, unii-hb95grm95d, m-carboxybenzaldehyde, hb95grm95d, 3-formylbenzoicacid, isophthalaldehydsaure
PubChem CID 12077
IUPAC Name 3-formylbenzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO2CC6H4CHO
Quantity 5g
Formula Weight 150.13
Percent Purity 98%
Chemical Name or Material 3-Carboxybenzaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-585-6

RUO – Research Use Only