missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxy-4-methoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog No. AAH5300503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5300503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5300503 Supplier Thermo Scientific Chemicals Supplier No. H5300503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 913836-12-1
Molecular Formula C8H9BO5
Molecular Weight (g/mol) 195.965
MDL Number MFCD06203322
InChI Key YZKWFWNYFKBAHO-UHFFFAOYSA-N
Synonym 3-carboxy-4-methoxyphenylboronic acid, 5-dihydroxyboranyl-2-methoxybenzoic acid, benzoicacid, 5-borono-2-methoxy-9ci, 3-carboxy-4-methoxybenzeneboronic acid, benzoic acid, 5-borono-2-methoxy, acmc-209raw, ksc486i0n, 5-dihydroxyboryl-2-methoxybenzoic acid
PubChem CID 44119154
IUPAC Name 5-borono-2-methoxybenzoic acid
SMILES B(C1=CC(=C(C=C1)OC)C(=O)O)(O)O

Specifications

Melting Point 176°C to 177°C
Quantity 1 g
Formula Weight 195.97
Percent Purity 98%
Chemical Name or Material 3-Carboxy-4-methoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only