missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxy-1-(4-sulfophenyl)-5-pyrazolone Sodium Salt 98.0+%, TCI America™

Catalog No. C084525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C084525G 25 g
C0845500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C084525G Supplier TCI America Supplier No. C084525G

Chemical Identifiers

CAS 52126-51-9
Molecular Formula C10H7N2NaO6S
Molecular Weight (g/mol) 306.224
MDL Number MFCD00059720
InChI Key KFVXGVBBXSOYNO-UHFFFAOYSA-M
Synonym 5-Oxo-1-(4-sulfophenyl)-2-pyrazoline-3-carboxylic Acid Sodium Salt
PubChem CID 23695324
IUPAC Name sodium;5-oxo-1-(4-sulfophenyl)-4H-pyrazole-3-carboxylate
SMILES C1C(=NN(C1=O)C2=CC=C(C=C2)S(=O)(=O)O)C(=O)[O-].[Na+]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 306.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Carboxy-1-(4-sulfophenyl)-5-pyrazolone Sodium Salt
EINECSNumber (5)-3450
TSCA Yes