missing translation for 'onlineSavingsMsg'
Learn More

3-Carbamyl-1-methylpyridinium Chloride 98.0+%, TCI America™

Catalog No. p-7132130
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03755G 5 g
M037525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M03755G Supplier TCI America Supplier No. M03755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1005-24-9 | C7H9ClN2O | 172.612 g/mol

Chemical Identifiers

CAS 1005-24-9
Molecular Formula C7H9ClN2O
Molecular Weight (g/mol) 172.612
MDL Number MFCD00060042
InChI Key BWVDQVQUNNBTLK-UHFFFAOYSA-N
Synonym 3-Carbamoyl-1-methylpyridinium Chloride
PubChem CID 70495
IUPAC Name 1-methylpyridin-1-ium-3-carboxamide;chloride
SMILES C[N+]1=CC=CC(=C1)C(=O)N.[Cl-]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 172.61
Percent Purity ≥98.0% (HPLC,T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material 3-Carbamyl-1-methylpyridinium Chloride
RTECSNumber UU1925000
TSCA No