missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Carbamoylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$212.51 - $212.51
Specifications
| CAS | 351422-73-6 |
|---|---|
| Melting Point | 226°C |
| Color | White-Yellow |
| Molecular Formula | C7H8BNO3 |
| MDL Number | MFCD03411948 |
Specifications
| 351422-73-6 | |
| White-Yellow | |
| MFCD03411948 | |
| (3-carbamoylphenyl)boronic acid | |
| 164.96 |
| 226°C | |
| C7H8BNO3 | |
| WDGWHKRJEBENCE-UHFFFAOYSA-N | |
| 2737814 | |
| Crystalline Powder |