missing translation for 'onlineSavingsMsg'
Learn More

3-Carbamoylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C29921G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C29921G Supplier TCI America Supplier No. C29921G

Specifications

Chemical Name or Material 3-Carbamoylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 351422-73-6
Melting Point 226°C
Color White-Yellow
Molecular Formula C7H8BNO3
Quantity 1 g
MDL Number MFCD03411948
Synonym 3-aminocarbonylphenylboronic acid, 3-carbamoylphenyl boronic acid, benzamide-3-boronic acid, 3-carbamoylbenzeneboronic acid, 3-aminocarbonyphenylboronic acid, 3-aminocarbonyl benzeneboronic acid, 3-aminocarbonylphenyl boronic acid, 3-carboxamidephenylboronic acid, 3-aminocarbonyl phenylboronic acid, 3-aminocarbonyl phenyl boronic acid
InChI Key WDGWHKRJEBENCE-UHFFFAOYSA-N
SMILES NC(=O)C1=CC=CC(=C1)B(O)O
IUPAC Name (3-carbamoylphenyl)boronic acid
Molecular Weight (g/mol) 164.96
PubChem CID 2737814
Formula Weight 164.96
Physical Form Crystalline Powder
Show More Show Less