missing translation for 'onlineSavingsMsg'
Learn More

3-Butenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

CAS: 7103-09-5 | C4H8O | 72.11 g/mol

$2853.32 - $2853.32

Chemical Identifiers

CAS 7103-09-5
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC427468000
SDS View Documents
Thermo Scientific Chemicals
427468000
800 mL AcroSeal™ Glass Bottle
Each for $2,853.32
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7103-09-5

Specifications

7103-09-5,109-99-9
6.8,89.8
0.9370g/mL
800 mL
AcroSeal™ Glass Bottle
0.937
H2C=CHCH2CH2MgBr
grignard reagent
Br[Mg]CCC=C
159.31
159.31
Brown to Gray
10.2,93.2
65°C
-30°C
Solution
C4H7BrMg
MFCD00209554
SMLLHQBZPPSOSR-UHFFFAOYSA-M
zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride
131675247
3-Butenylmagnesium bromide, 0.5M solution in THF
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.
Highly flammable liquid and vapor.
Suspected of causing cancer.
Reacts violently

GHS P Statement
Wear eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only