missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothiophene, 97%, Thermo Scientific™

Chemical Identifiers

CAS 872-31-1
Molecular Formula C4H3BrS
Molecular Weight (g/mol) 163.03
MDL Number MFCD00005464
InChI Key XCMISAPCWHTVNG-UHFFFAOYSA-N
Synonym 3-thienyl bromide, thiophene, 3-bromo, 3-bromo thiophene, beta-bromothiophene, .beta.-bromothiophene, 3-bromo-thiophene, bromothiophene 3-, 3-bromothiphene, 3-bromthiophene, pubchem5306
PubChem CID 13383
IUPAC Name 3-bromothiophene
SMILES BrC1=CSC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC147805000
SDS View Documents
Thermo Scientific Chemicals
147805000
500mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

872-31-1
163.03
XCMISAPCWHTVNG-UHFFFAOYSA-N
13383
BrC1=CSC=C1
C4H3BrS
MFCD00005464
3-thienyl bromide, thiophene, 3-bromo, 3-bromo thiophene, beta-bromothiophene, .beta.-bromothiophene, 3-bromo-thiophene, bromothiophene 3-, 3-bromothiphene, 3-bromthiophene, pubchem5306
3-bromothiophene

Specifications

872-31-1
158.0°C
Authentic
Glass bottle
1.5905 to 1.5925
500mL
3-thienyl bromide, thiophene, 3-bromo, 3-bromo thiophene, beta-bromothiophene, .beta.-bromothiophene, 3-bromo-thiophene, bromothiophene 3-, 3-bromothiphene, 3-bromthiophene, pubchem5306
XCMISAPCWHTVNG-UHFFFAOYSA-N
3-bromothiophene
13383
97%
1.7400g/mL
56°C
96% min. (GC)
C4H3BrS
MFCD00005464
1.74
Solubility in water: immiscible. Other solubilities: soluble in acetone,benzene,slightly soluble in chloroform
BrC1=CSC=C1
163.03
163.03
3-Bromothiophene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal in contact with skin.
Causes skin irritation.
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irr

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly

GHS Signal Word: Danger

EINECSNumber : 212-821-3

Recommended Storage : Product may darken on storage

SDS
Documents

Documents

RUO – Research Use Only