missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothiophene-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. B46051G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46051G 1 g
B46055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46051G Supplier TCI America Supplier No. B46051G

Chemical Identifiers

CAS 7311-64-0
Molecular Formula C5H3BrO2S
Molecular Weight (g/mol) 207.041
MDL Number MFCD00052291
InChI Key VQQLWBUTTMNMFT-UHFFFAOYSA-N
Synonym 3-bromo-2-thiophenecarboxylic acid, 3-bromo-2-thenoic acid, 2-thiophenecarboxylic acid, 3-bromo, 3-bromo-thiophene-2-carboxylic acid, maybridge1_007344, 3-bromothiophene-2-carboxylicacid, bromothiophenecarboxylic, pubchem10096, ksc494c9j, bromo-thiophene-2-carboxylic acid
PubChem CID 2739688
IUPAC Name 3-bromothiophene-2-carboxylic acid
SMILES C1=CSC(=C1Br)C(=O)O

Specifications

Melting Point 198°C
Color White-Yellow
Quantity 1 g
Formula Weight 207.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromothiophene-2-carboxylic Acid