missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothiophene-2-carboxylic acid, 97%

Catalog No. AAA1193503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1193503 1 g
AAA1193506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1193503 Supplier Thermo Scientific Chemicals Supplier No. A1193503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7311-64-0 | C5H3BrO2S | 207.041 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7311-64-0
Molecular Formula C5H3BrO2S
Molecular Weight (g/mol) 207.041
MDL Number MFCD00052291
InChI Key VQQLWBUTTMNMFT-UHFFFAOYSA-N
Synonym 3-bromo-2-thiophenecarboxylic acid, 3-bromo-2-thenoic acid, 2-thiophenecarboxylic acid, 3-bromo, 3-bromo-thiophene-2-carboxylic acid, maybridge1_007344, 3-bromothiophene-2-carboxylicacid, bromothiophenecarboxylic, pubchem10096, ksc494c9j, bromo-thiophene-2-carboxylic acid
PubChem CID 2739688
IUPAC Name 3-bromothiophene-2-carboxylic acid
SMILES C1=CSC(=C1Br)C(=O)O

Specifications

Melting Point 195°C to 196°C
Quantity 1 g
Beilstein 121585
Formula Weight 207.05
Percent Purity 97%
Chemical Name or Material 3-Bromothiophene-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only