missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothiophene-2-carboxamide, 99%, Thermo Scientific Chemicals

Catalog No. aal1613803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1613803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1613803 Supplier Thermo Scientific Chemicals Supplier No. L1613803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 78031-18-2 | Molecular Formula: C5H4BrNOS | Molecular Weight: 206.06 g/mol

3-Bromothiophene-2-carboxamide is used in Stille cross-coupling conditions were used to couple the resin-bound peptide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromothiophene-2-carboxamide is used in Stille cross-coupling conditions were used to couple the resin-bound peptide.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 78031-18-2
Molecular Formula C5H4BrNOS
Molecular Weight (g/mol) 206.06
MDL Number MFCD00173835
InChI Key KWXUWNXTRUZHNQ-UHFFFAOYSA-N
Synonym maybridge1_007895, 3-bromo-2-carboxamidothiophene, 2-thiophenecarboxamide,3-bromo, 3-bromo-thiophene-2-carboxamide, 3-bromo-2-thiophenecarboxamide #, buttpark 180\02-08, 3-bromo thiophene-2-carboxamide
PubChem CID 605721
IUPAC Name 3-bromothiophene-2-carboxamide
SMILES NC(=O)C1=C(Br)C=CS1

Specifications

Melting Point 106°C to 108°C
Quantity 1 g
Beilstein 4937660
Solubility Information Insoluble in water.
Formula Weight 206.06
Percent Purity 99%
Chemical Name or Material 3-Bromothiophene-2-carboxamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only