missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Bromoquinoline 98.0+%, TCI America™
Manufacturer: Tci America B101325G
Specifications
3-Bromoquinoline | |
Yellow | |
25g | |
C9H6BrN | |
3-bromo quinoline, 3-bromo-quinoline, 3-bromoquinliine, 3-bromoquinolin, 3-bromoquinoline, acmc-1ax3i, pubchem5814, quinoline, 3-bromo | |
C1=CC=C2C(=C1)C=C(C=N2)Br | |
208.058 | |
208.06 | |
≥98.0% (GC,T) |
276°C | |
14°C | |
5332-24-1 | |
MFCD00006767 | |
ZGIKWINFUGEQEO-UHFFFAOYSA-N | |
3-bromoquinoline | |
21413 | |
Clear Liquid at 20°C |
Chemical Identifiers
5332-24-1 | |
208.058 | |
ZGIKWINFUGEQEO-UHFFFAOYSA-N | |
21413 | |
C1=CC=C2C(=C1)C=C(C=N2)Br |
C9H6BrN | |
MFCD00006767 | |
3-bromo quinoline, 3-bromo-quinoline, 3-bromoquinliine, 3-bromoquinolin, 3-bromoquinoline, acmc-1ax3i, pubchem5814, quinoline, 3-bromo | |
3-bromoquinoline |
Safety and Handling
TSCA : Yes