missing translation for 'onlineSavingsMsg'
Learn More

3-Bromoquinoline, 98%

Catalog No. p-7022001
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
100 g
250 g
50 g
Catalog No. Quantity
AAA1457309 10 g
AAA1457318 50 g
AAA1457322 100 g
AAA1457330 250 g
4 options

Catalog No. AAA1457309

Supplier: Thermo Scientific Chemicals A1457309

Only null left

CAS: 5332-24-1 | C9H6BrN | 208.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5332-24-1
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.058
MDL Number MFCD00006767
InChI Key ZGIKWINFUGEQEO-UHFFFAOYSA-N
Synonym quinoline, 3-bromo, 3-bromoquinliine, 3-bromo-quinoline, 3-bromo quinoline, 3-bromoquinolin, pubchem5814, 3-bromoquinoline, acmc-1ax3i
PubChem CID 21413
IUPAC Name 3-bromoquinoline
SMILES C1=CC=C2C(=C1)C=C(C=N2)Br

Specifications

Melting Point 13°C to 15°C
Density 1.533
Boiling Point 274°C to 276°C
Flash Point >110°C (230°F)
Refractive Index 1.664
Quantity 10 g
Beilstein 112939
Formula Weight 208.06
Percent Purity 98%
Chemical Name or Material 3-Bromoquinoline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 226-237-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only