missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopyruvic acid hydrate, 98%

Catalog No. AC325690250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC325690250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC325690250 Supplier Thermo Scientific Chemicals Supplier No. 325690250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 206860-50-6 | C3H3BrO3 | 166.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

CAS 206860-50-6, 1113-59-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 79°C to 82°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C3H3BrO3
Quantity 25 g
MDL Number MFCD00149107
Synonym 3-bromopyruvic acid hydrate, 3-bromo-2-oxopropanoic acid hydrate, bromopyruvic acid hydrate, 3-bromo-2-oxopropanoic acid, hydrate, 3-bromo-2-oxopropanoic acid-water 1/1, propanoic acid, 3-bromo-2-oxo-, monohydrate 9ci
Solubility Information Solubility in water: soluble
InChI Key PRRZDZJYSJLDBS-UHFFFAOYSA-N
SMILES OC(=O)C(=O)CBr
IUPAC Name 3-bromo-2-oxopropanoic acid;hydrate
Molecular Weight (g/mol) 166.96
PubChem CID 44118703
Formula Weight 166.96
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Bromopyruvic acid hydrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa
EINECSNumber 214-206-5
RTECSNumber UZ0840000
TSCA TSCA

RUO – Research Use Only