Learn More
3-Bromopyrrolidine hydrobromide, 96%, Thermo Scientific Chemicals
$195.95 - $195.95
Chemical Identifiers
| CAS | 1262769-75-4 |
|---|---|
| Molecular Formula | C4H9Br2N |
| Molecular Weight (g/mol) | 230.931 |
| MDL Number | MFCD13181592 |
| InChI Key | ZSDJSWROXIUGAE-UHFFFAOYSA-N |
| Synonym | 3-bromopyrrolidine hydrobromide, 3-bromopyrrolidine hbr, 3-bromopyrrolidine-hydrogen bromide 1/1 |
| PubChem CID | 66570691 |
| IUPAC Name | 3-bromopyrrolidine;hydrobromide |
| SMILES | C1CNCC1Br.Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6683303
|
Thermo Scientific Chemicals
H6683303 |
1 g |
Each for $195.95
|
|
|||||
Description
It is employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a intermediate for pharmaceutical.
Notes
Moisture and Light Sensitive, store in dark and away form water/moisture. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
| 1262769-75-4 | |
| 230.931 | |
| ZSDJSWROXIUGAE-UHFFFAOYSA-N | |
| 66570691 | |
| C1CNCC1Br.Br |
| C4H9Br2N | |
| MFCD13181592 | |
| 3-bromopyrrolidine hydrobromide, 3-bromopyrrolidine hbr, 3-bromopyrrolidine-hydrogen bromide 1/1 | |
| 3-bromopyrrolidine;hydrobromide |
Specifications
| 1262769-75-4 | |
| Brown to Pink | |
| MFCD13181592 | |
| UN3335 | |
| 3-bromopyrrolidine hydrobromide, 3-bromopyrrolidine hbr, 3-bromopyrrolidine-hydrogen bromide 1/1 | |
| C1CNCC1Br.Br | |
| 230.931 | |
| 230.93 | |
| Solid |
| 135°C to 141°C | |
| C4H9Br2N | |
| 1 g | |
| Light and moisture sensitive | |
| ZSDJSWROXIUGAE-UHFFFAOYSA-N | |
| 3-bromopyrrolidine;hydrobromide | |
| 66570691 | |
| 96% | |
| 3-Bromopyrrolidine hydrobromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only