missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopyrrolidine hydrobromide, 96%, Thermo Scientific Chemicals

Catalog No. AAH6683303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6683303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6683303 Supplier Thermo Scientific Chemicals Supplier No. H6683303

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Notes
Moisture and Light Sensitive, store in dark and away form water/moisture. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 1262769-75-4
Molecular Formula C4H9Br2N
Molecular Weight (g/mol) 230.931
MDL Number MFCD13181592
InChI Key ZSDJSWROXIUGAE-UHFFFAOYSA-N
Synonym 3-bromopyrrolidine hydrobromide, 3-bromopyrrolidine hbr, 3-bromopyrrolidine-hydrogen bromide 1/1
PubChem CID 66570691
IUPAC Name 3-bromopyrrolidine;hydrobromide
SMILES C1CNCC1Br.Br

Specifications

Melting Point 135°C to 141°C
Color Brown to Pink
Quantity 1 g
UN Number UN3335
Sensitivity Light and moisture sensitive
Formula Weight 230.93
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Bromopyrrolidine hydrobromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only