missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopyridine-2-carboxylic acid, 97%

CAS: 30683-23-9 | C6H4BrNO2 | 202.007 g/mol

Supplier:  Thermo Scientific Chemicals H6425814

 View more versions of this product

Catalog No. AAH6425814


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Bromopyridine-2-carboxylic acid
141°C to 144°C
C6H4BrNO2
25 g
3-bromopicolinic acid, 3-bromo-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 3-bromo, 3-bromopyridine-2-carboxylicacid, 3-bromo-2-pyridine carboxylic acid, 3-bromo-pyridine-2-carboxylic acid, pubchem5070, bromopyridine carboxylic, acmc-1cpch, 3-bromo-2-picolinic acid
C1=CC(=C(N=C1)C(=O)O)Br
202.007
202.01
Solid
30683-23-9
1.813 g/mL
MFCD01320380
472171
KBDIRPOTVAODSA-UHFFFAOYSA-N
3-bromopyridine-2-carboxylic acid
2050129
97%

Chemical Identifiers

30683-23-9
202.007
KBDIRPOTVAODSA-UHFFFAOYSA-N
2050129
C1=CC(=C(N=C1)C(=O)O)Br
C6H4BrNO2
MFCD01320380
3-bromopicolinic acid, 3-bromo-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 3-bromo, 3-bromopyridine-2-carboxylicacid, 3-bromo-2-pyridine carboxylic acid, 3-bromo-pyridine-2-carboxylic acid, pubchem5070, bromopyridine carboxylic, acmc-1cpch, 3-bromo-2-picolinic acid
3-bromopyridine-2-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only