missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropylboronic acid pinacol ester, 98%

Catalog No. AAH3064606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3064606 5 g
AAH3064603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3064606 Supplier Thermo Scientific Chemicals Supplier No. H3064606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 124215-44-7 | C9H18BBrO2 | 248.96 g/mol

Used as primary and secondary intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as primary and secondary intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 124215-44-7
Molecular Formula C9H18BBrO2
Molecular Weight (g/mol) 248.96
MDL Number MFCD10567053
InChI Key CHQXFJUKMDJWHO-UHFFFAOYSA-N
Synonym 2-3-bromopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-bromopropylboronic acid pinacol ester, 1,3,2-dioxaborolane,2-3-bromopropyl-4,4,5,5-tetramethyl, acmc-1c5j4, 3-bromopropylboronic acid, pinacol ester, 2-3-bromopropyl-4?4?5?5-tetramethyl-1?3?2-dioxaborolane
PubChem CID 14836669
IUPAC Name 2-(3-bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(CCCBr)OC1(C)C

Specifications

Density 1.177
Boiling Point 61°C to 64°C (0.35 mmHg)
Quantity 5 g
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 248.96
Percent Purity 98%
Chemical Name or Material 3-Bromopropylboronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No

RUO – Research Use Only