missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropylamine Hydrobromide 98.0+%, TCI America™

Catalog No. B126325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B126325G 25 g
B1263100G 100 g
B1263500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B126325G Supplier TCI America Supplier No. B126325G

Chemical Identifiers

CAS 5003-71-4
Molecular Formula C3H9Br2N
Molecular Weight (g/mol) 218.92
MDL Number MFCD00012912
InChI Key PQIYSSSTRHVOBW-UHFFFAOYSA-N
Synonym 3-bromopropylamine hydrobromide, 3-bromopropan-1-amine hydrobromide, 3-aminopropyl bromide hydrobromide, 1-propanamine, 3-bromo-, hydrobromide, 3-bromopropylammonium bromide, 3-bromopropanamine hydrobromide, 1-amino-3-bromopropane hydrobromide, 3-bromo-1-propanamine, hydrobromide, 3-brominepropylaminehydrobromide, 3-bromopropylamine, hydrobromide
PubChem CID 78701
IUPAC Name 3-bromopropan-1-amine;hydrobromide
SMILES C(CN)CBr.Br

Specifications

Melting Point 174°C
Color White
Quantity 25 g
Formula Weight 218.92
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromopropylamine Hydrobromide
RTECSNumber UH9925000
TSCA Yes