missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Bromopropylamine hydrobromide, 98%, ACROS Organics™
$60.20 - $60.20
Chemical Identifiers
CAS | 5003-71-4 |
---|---|
Molecular Formula | C3H8BrN·HBr |
Molecular Weight (g/mol) | 218.93 |
MDL Number | MFCD00012912 |
InChI Key | PQIYSSSTRHVOBW-UHFFFAOYSA-N |
Synonym | 1-amino-3-bromopropane hydrobromide, 1-propanamine, 3-bromo-, hydrobromide, 3-aminopropyl bromide hydrobromide, 3-brominepropylaminehydrobromide, 3-bromo-1-propanamine, hydrobromide, 3-bromopropan-1-amine hydrobromide, 3-bromopropanamine hydrobromide, 3-bromopropylamine hydrobromide, 3-bromopropylamine, hydrobromide, 3-bromopropylammonium bromide |
PubChem CID | 78701 |
IUPAC Name | 3-bromopropan-1-amine;hydrobromide |
SMILES | C(CN)CBr.Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC107380250
|
Fisher Scientific Acros
107380250 |
25g | Glass bottle |
Each for $60.20
|
|
AC107381000
|
Fisher Scientific Acros
107381000 |
100g | Glass bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
5003-71-4 | |
218.93 | |
PQIYSSSTRHVOBW-UHFFFAOYSA-N | |
78701 | |
C(CN)CBr.Br |
C3H8BrN·HBr | |
MFCD00012912 | |
1-amino-3-bromopropane hydrobromide, 1-propanamine, 3-bromo-, hydrobromide, 3-aminopropyl bromide hydrobromide, 3-brominepropylaminehydrobromide, 3-bromo-1-propanamine, hydrobromide, 3-bromopropan-1-amine hydrobromide, 3-bromopropanamine hydrobromide, 3-bromopropylamine hydrobromide, 3-bromopropylamine, hydrobromide, 3-bromopropylammonium bromide | |
3-bromopropan-1-amine;hydrobromide |
Specifications
Authentic | |
White | |
25g | |
97.5 | |
98% | |
Br(CH2)3NH2·HBr | |
04, 149 | |
Solubility in water: soluble | |
C(CN)CBr.Br | |
218.93 | |
218.93 | |
98% |
Glass bottle | |
171.0°C to 175.0°C | |
5003-71-4 | |
100.0 | |
C3H8BrN·HBr | |
MFCD00012912 | |
1-amino-3-bromopropane hydrobromide, 1-propanamine, 3-bromo-, hydrobromide, 3-aminopropyl bromide hydrobromide, 3-brominepropylaminehydrobromide, 3-bromo-1-propanamine, hydrobromide, 3-bromopropan-1-amine hydrobromide, 3-bromopropanamine hydrobromide, 3-bromopropylamine hydrobromide, 3-bromopropylamine, hydrobromide, 3-bromopropylammonium bromide | |
PQIYSSSTRHVOBW-UHFFFAOYSA-N | |
3-bromopropan-1-amine;hydrobromide | |
78701 | |
Powder or Chunks | |
3-Bromopropylamine hydrobromide, 98% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 225-675-0
RTECSNumber : UH9925000
TSCA : TSCA