missing translation for 'onlineSavingsMsg'
Learn More

(3-Bromopropyl)trimethoxysilane 98.0+%, TCI America™

Catalog No. p-7122852
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32895G 5 g
B328925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32895G Supplier TCI America Supplier No. B32895G

Chemical Identifiers

CAS 51826-90-5
Molecular Formula C6H15BrO3Si
Molecular Weight (g/mol) 243.172
MDL Number MFCD00053894
InChI Key GLISZRPOUBOZDL-UHFFFAOYSA-N
PubChem CID 103984
IUPAC Name 3-bromopropyl(trimethoxy)silane
SMILES CO[Si](CCCBr)(OC)OC

Specifications

Color Yellow
Boiling Point 130°C
Quantity 5 g
Formula Weight 243.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (3-Bromopropyl)trimethoxysilane
EINECSNumber (2)-2079
TSCA No