missing translation for 'onlineSavingsMsg'
Learn More

(3-Bromopropyl)phosphonic Acid 98.0+%, TCI America™

Catalog No. p-7119139
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15911G 1 g
B15915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15911G Supplier TCI America Supplier No. B15911G
Only null left

Chemical Identifiers

CAS 1190-09-6
Molecular Formula C3H8BrO3P
Molecular Weight (g/mol) 202.972
MDL Number MFCD00191385
InChI Key CXOIECRVHUDBSA-UHFFFAOYSA-N
Synonym 3-Bromopropanephosphonic Acid
PubChem CID 4376761
IUPAC Name 3-bromopropylphosphonic acid
SMILES C(CP(=O)(O)O)CBr

Specifications

Melting Point 112°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 202.97
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (3-Bromopropyl)phosphonic Acid