missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Bromopropyl)phosphonic Acid 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 112°C |
| Color | White-Yellow |
| Quantity | 1 g |
| UN Number | 3261 |
| Formula Weight | 202.97 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (3-Bromopropyl)phosphonic Acid |
Sécurité et manipulation
| TSCA | No |