missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromopropyl Acetate 95.0+%, TCI America™
$109.98 - $380.27
Chemical Identifiers
| CAS | 592-33-6 |
|---|---|
| Molecular Formula | C5H9BrO2 |
| MDL Number | MFCD02258489 |
| Synonym | Acetic Acid 3-Bromopropyl Ester |
Chemical Identifiers
| 592-33-6 | |
| C5H9BrO2 | |
| MFCD02258489 | |
| Acetic Acid 3-Bromopropyl Ester |
Specifications
| 592-33-6 | |
| C5H9BrO2 | |
| 1 g | |
| 181.03 | |
| Liquid |
| 163°C | |
| MFCD02258489 | |
| Acetic Acid 3-Bromopropyl Ester | |
| ≥98.0% (GC) | |
| 3-Bromopropyl Acetate |
Safety and Handling
TSCA : No