missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromopropyl Acetate 95.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 163°C |
| Quantity | 5 g |
| Formula Weight | 181.03 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Bromopropyl Acetate |
Safety and Handling
| TSCA | No |